Learn More
Abcam Gentamycin sulfate (aqueous), aminoglycoside antibiotic agent, 10ML
Catalog No. 502704568
Encompass_Preferred
MW 0 g/mol. Broad spectrum aminoglycoside antibiotic active against many strains of gram-negative bacteria, the gram-positive Staphylococcus aeruginosa as well as some strains of Mycobacterium tuberculosis. Inhibits protein synthesis in susceptible bacteria, by binding to the L6 protein of the 50S ribosomal subunit.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4568 Supplier Abcam Supplier No. AB14657310ML
MW 0 g/mol. Broad spectrum aminoglycoside antibiotic active against many strains of gram-negative bacteria, the gram-positive Staphylococcus aeruginosa as well as some strains of Mycobacterium tuberculosis. Inhibits protein synthesis in susceptible bacteria, by binding to the L6 protein of the 50S ribosomal subunit.
The product is subject to the following: Abcam Restricted Use Statement