Learn More
AG Scientific Inc Gentamycin Sulfate Sol 5X10ML
Catalog No. 502268299
Encompass_Preferred
Gentamycin Sulfate sterile filtrate solution is a broad spectrum, cell culture antibiotic that is non toxic to viruses and mammalian cells at antibacterial levels. It is useful for long-term virus and tissue culture studies.
Catalog No. 50-226-8299 Supplier AG Scientific Inc Supplier No. G1068505X10ML
Gentamycin Sulfate sterile filtrate solution is a broad spectrum, cell culture antibiotic that is non toxic to viruses and mammalian cells at antibacterial levels. It is useful for long-term virus and tissue culture studies.