Learn More
Gentamycin Sulfate Solution (10mg Gentamycin Base Per ML), MP Biomedicals
Shop All MP Biomedicals, Inc ProductsSpecifications
Specifications
| Type | Gentamicin Sulfate |
| Physical Form | Liquid |
| Color | Colorless |
| Quantity | 10 x 10 mL |
| Grade | Not Available |
| Chemical Name or Material | Gentamycin Sulfate Solution |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.