Learn More
Med Vet International First Aid Antibiotic Humco™ Topical Liquid 2 oz. Bottle, 00395100392, Each
Catalog No. 503044398
Encompass_Preferred
1% gentian violet, ethyl alcohol 10% (preservative) purified water Humco's gentian violet is for the treatment and infection-prevention of minor cuts and scrapes Gentian violet is an excellent treatment for Candida albicans Gentian violet is also a dye that is commonly used in school labs as the basis of the gram stain for detecting the presence of Gram-positive bacteria 2-ounce bottle
Catalog No. 50-304-4398 Supplier Med Vet International Supplier No. 713309EA
1% gentian violet, ethyl alcohol 10% (preservative) purified water Humco's gentian violet is for the treatment and infection-prevention of minor cuts and scrapes Gentian violet is an excellent treatment for Candida albicans Gentian violet is also a dye that is commonly used in school labs as the basis of the gram stain for detecting the presence of Gram-positive bacteria 2-ounce bottle