Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentian Violet, 1% Alcoholic, Certified, LabChem™

Catalog No. LC148001 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC148001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC148001 Supplier LabChem, Inc. Supplier No. LC148001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory liquid solution for staining applications, supplied in a 500 mL poly bottle.

Specifications

Chemical Name or Material Gentian Violet
Name Note 1% Alcoholic
CAS Max % 89.1, 4.95, 4.95, 1
CAS 67-63-0, 67-56-1, 548-62-9
Identification Passes Test
Packaging Poly Bottle
Suitability Passes Test
Linear Formula C25H30CIN3
Decomposition Information Fume; Carbon monoxide; Carbon dioxide; May release flammable gases
Color Purple
Solubility Information Soluble in water
Quantity 500 mL
Physical Form Liquid
Molecular Weight (g/mol) 407.99
Formula Weight 407.99
Grade Certified
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.