Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentian Violet, Hucker Formulation, for Gram Staining, Ricca Chemical
SDP

Catalog No. 32404 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3240-4 120 mL Natural Poly Bottle
3240-16 500 mL Natural Poly Bottle
3240-32 1 L Natural Poly Bottle
3240-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 3240-4 Supplier Ricca Chemical Company Supplier No. 32404
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Gram Staining Technique

Specifications

Chemical Name or Material Gentian Violet
Name Note Hucker Formulation
CAS 7732-18-5, 64-17-5, 548-62-9, 6009-70-7, 64-56-1
CAS Min % 82.0, 13.2, 1.65, 0.66
CAS Max % 86.0, 13.74, 1.72, 0.69
Packaging Natural Poly Bottle
Color Blue/Purple
Quantity 120 mL
Physical Form Liquid
Grade Indicator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.