Learn More
Medchemexpress LLC Gentiopicroside | 20831-76-9 | 356.32 | 25 MG
SDP
Catalog No. 5000389911
Encompass_Preferred
Gentiopicroside is a naturally occurring iridoid glycoside that inhibits P450 activity and exhibits anti-inflammatory and antioxidative effects.
  • Naturally occurring iridoid glycoside
  • Inhibits P450 activity
  • Has anti-inflammatory effects
  • Has antioxidative effects
  • Inhibits RANKL-induced osteoclast formation
  • Blocks expression of osteoclast-related proteins
  • Prevents RANKL-triggered JNK and NF-κB activation
  • Inhibits RANKL-induced bone resorption
  • Reduces gastric ulcer index (in vivo)
Catalog No. 50-003-89911 Supplier Medchemexpress LLC Supplier No. 25MGHYN0494
Gentiopicroside is a naturally occurring iridoid glycoside that inhibits P450 activity and exhibits anti-inflammatory and antioxidative effects.
  • Naturally occurring iridoid glycoside
  • Inhibits P450 activity
  • Has anti-inflammatory effects
  • Has antioxidative effects
  • Inhibits RANKL-induced osteoclast formation
  • Blocks expression of osteoclast-related proteins
  • Prevents RANKL-triggered JNK and NF-κB activation
  • Inhibits RANKL-induced bone resorption
  • Reduces gastric ulcer index (in vivo)