Learn More
TARGETMOL CHEMICALS INC GENTIOPICROSIDE 25MG
SDP
Catalog No. 502577048
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Gentiopicroside (Gentiopicrin) a naturally occurring iridoid glycoside extracted from Gentiana manshurica Kitag inhibits P450 activity with an IC50 and a Ki of 61 uM and 22.8 uM for CYP2A6; has antianti-inflammatoryand antioxidative effects. purity: 99%
Catalog No. 50-257-7048 Supplier TARGETMOL CHEMICALS INC Supplier No. T277025MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Gentiopicroside (Gentiopicrin) a naturally occurring iridoid glycoside extracted from Gentiana manshurica Kitag inhibits P450 activity with an IC50 and a Ki of 61 uM and 22.8 uM for CYP2A6; has antianti-inflammatoryand antioxidative effects. purity: 99%