Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gentisic acid sodium salt hydrate

Catalog No. G512950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G512950G 50 g
G512910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G512950G Supplier Sigma Aldrich Supplier No. G512950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant involved in oxidative coupling reactions followed by the product use as an antioxidant and tyrosinase inhibitor

Specifications

CAS 4955-90-2
Molecular Formula C7H5NaO4
Linear Formula C7H5O4Na
MDL Number MFCD00150273
Quantity 50 g
Synonym 2,5-Dihydroxybenzoic acid sodium salt; 5-Hydroxysalicylate sodium
Molecular Weight (g/mol) 176.1
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.