Learn More
Medchemexpress LLC Gentisin | 437-50-3 | 1 MG
SDP
Catalog No. 5000418552
Encompass_Preferred
Gentisin is a natural compound extracted from Gentianae radix (Gentianaceae). It exhibits mutagenic activity and can be utilized in vascular smooth muscle research. It inhibits PDGF-induced vascular smooth muscle cell proliferation in a concentration-dependent manner with an IC50 of 7.84 μM. It also shows cytotoxicity against Taxol-resistant human A549 cells (IC50: 103.76 μM), human MCF7/ADR cells (IC50: 31.82 μM), and Taxol-resistant human SMMC7721 cells (IC50: 87.44 μM) after 48 hours as measured by MTT assay.
  • For research use only.
  • Available in multiple sizes.
  • In stock for 1 mg size.
  • Data Sheet, COA, SDS, handling instructions, and technical support available.
  • In vivo dissolution calculator and Help & FAQs available.
  • Related screening libraries available.
  • Related small molecules listed.
Catalog No. 50-004-18552 Supplier Medchemexpress LLC Supplier No. HYN42791MG
Gentisin is a natural compound extracted from Gentianae radix (Gentianaceae). It exhibits mutagenic activity and can be utilized in vascular smooth muscle research. It inhibits PDGF-induced vascular smooth muscle cell proliferation in a concentration-dependent manner with an IC50 of 7.84 μM. It also shows cytotoxicity against Taxol-resistant human A549 cells (IC50: 103.76 μM), human MCF7/ADR cells (IC50: 31.82 μM), and Taxol-resistant human SMMC7721 cells (IC50: 87.44 μM) after 48 hours as measured by MTT assay.
  • For research use only.
  • Available in multiple sizes.
  • In stock for 1 mg size.
  • Data Sheet, COA, SDS, handling instructions, and technical support available.
  • In vivo dissolution calculator and Help & FAQs available.
  • Related screening libraries available.
  • Related small molecules listed.