Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gentisyl alcohol | 495-08-9 | MFCD06202616 | 98.9% | 140.14 | C7H8O3 | 25 MG
SDP

Catalog No. 5000280854
Encompass_Preferred

Gentisyl alcohol is a phenolic compound (2-(hydroxymethyl)benzene-1,4-diol) used as a research reagent with reported activity against Staphylococcus aureus. It is supplied as a high-purity solid for laboratory research and biochemical studies. The material is intended for research use only and is provided with supporting documentation.

  • High purity: 98.9%.
  • Molecular formula: C7H8O3.
  • Molecular weight: 140.14.
  • Supplied in small research sizes, including 25 mg.
  • Provided with datasheet and certificate of analysis.
  • Intended for antibacterial and biochemical research applications.

Catalog No. 50-002-80854 Supplier Medchemexpress LLC Supplier No. HYW24890725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gentisyl alcohol is a phenolic compound (2-(hydroxymethyl)benzene-1,4-diol) used as a research reagent with reported activity against Staphylococcus aureus. It is supplied as a high-purity solid for laboratory research and biochemical studies. The material is intended for research use only and is provided with supporting documentation.

  • High purity: 98.9%.
  • Molecular formula: C7H8O3.
  • Molecular weight: 140.14.
  • Supplied in small research sizes, including 25 mg.
  • Provided with datasheet and certificate of analysis.
  • Intended for antibacterial and biochemical research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.