Learn More
Medchemexpress LLC Gentisyl alcohol | 495-08-9 | MFCD06202616 | 98.9% | 140.14 g/mol | C7H8O3 | 250 MG
SDP
Catalog No. 5000281289
Encompass_Preferred
Gentisyl alcohol is a research-grade small molecule reported to have antibacterial activity against Staphylococcus aureus. It is identified by CAS 495-08-9, has molecular formula C7H8O3, and molecular weight 140.14 g/mol. The compound is supplied in multiple pack sizes and is accompanied by product documentation for laboratory use.
  • High purity (98.9%).
  • Molecular formula C7H8O3; molecular weight 140.14 g/mol.
  • Reported anti-Staphylococcus aureus activity.
  • Available pack sizes include 25 mg, 50 mg, 100 mg, 250 mg, 500 mg, and 1 g.
  • Datasheet, certificate of analysis, and safety data sheet available.
Catalog No. 50-002-81289 Supplier Medchemexpress LLC Supplier No. HYW248907250MG
Gentisyl alcohol is a research-grade small molecule reported to have antibacterial activity against Staphylococcus aureus. It is identified by CAS 495-08-9, has molecular formula C7H8O3, and molecular weight 140.14 g/mol. The compound is supplied in multiple pack sizes and is accompanied by product documentation for laboratory use.
  • High purity (98.9%).
  • Molecular formula C7H8O3; molecular weight 140.14 g/mol.
  • Reported anti-Staphylococcus aureus activity.
  • Available pack sizes include 25 mg, 50 mg, 100 mg, 250 mg, 500 mg, and 1 g.
  • Datasheet, certificate of analysis, and safety data sheet available.