Learn More
STEMCELL Technologies Gentle Cell Dissociation Reagent, Size: 500 mL
SDP
Catalog No. 502390198
Encompass
Gentle Cell Dissociation Reagent (GCDR) is an enzyme-free reagent suitable for the dissociation of human embryonic stem (ES) cells or human induced pluripotent stem (iPS) cells into cell aggregates for routine passaging or into a single-cell suspension. It is also suitable for the isolation of intestinal crypts to establish intestinal organoids and for breaking up Corning Matrigel domes when passaging organoid cultures. GCDR does not contain enzymes or other proteins.
Catalog No. 50-239-0198 Supplier STEMCELL Technologies Supplier No. 1001077
Gentle Cell Dissociation Reagent (GCDR) is an enzyme-free reagent suitable for the dissociation of human embryonic stem (ES) cells or human induced pluripotent stem (iPS) cells into cell aggregates for routine passaging or into a single-cell suspension. It is also suitable for the isolation of intestinal crypts to establish intestinal organoids and for breaking up Corning Matrigel domes when passaging organoid cultures. GCDR does not contain enzymes or other proteins.