Learn More
STEMCELL Technologies Gentle Collagenase/Hyaluronidase, Size: 10 mL
SDP
Supplier:  STEMCELL Technologies 07919
Encompass
10X Collagenase/Hyaluronidase in Dulbecco’s Modified Eagle’s Medium (DMEM) for the gentle enzymatic dissociation of mouse mammary tissue.
Catalog No. NC2031809