Learn More
MSC Zoëcon Insecticide, Emulsified Concentrate, 16 oz, with IGR
Catalog No. 502839283
Encompass
Gentrol Complete EC3 provides broad-spectrum relief from kitchen insects and breaks the life cycles of German cockroaches, stored product pests, plus fruit flies and drain flies. It combines an insecticide with an insect growth regulator, designed for use in food handling establishments, and features a multi-rate formulation to tackle various levels of infestation.
  • Emulsified concentrate formulation that kills adult insects on contact.
  • Active ingredient, (S)-hydroprene, breaks the life cycle of kitchen bugs and prevents rebound infestations.
  • Approved for use in restaurant pest control, and treatments in other food and non-food handling establishments.
  • Multi-rate formulation to tackle all levels of a kitchen infestation.
  • Insect growth regulator translocates deep into wall cavities, cracks and crevices.
Catalog No. 50-283-9283 Supplier MSC Supplier No. 74334160
Gentrol Complete EC3 provides broad-spectrum relief from kitchen insects and breaks the life cycles of German cockroaches, stored product pests, plus fruit flies and drain flies. It combines an insecticide with an insect growth regulator, designed for use in food handling establishments, and features a multi-rate formulation to tackle various levels of infestation.
  • Emulsified concentrate formulation that kills adult insects on contact.
  • Active ingredient, (S)-hydroprene, breaks the life cycle of kitchen bugs and prevents rebound infestations.
  • Approved for use in restaurant pest control, and treatments in other food and non-food handling establishments.
  • Multi-rate formulation to tackle all levels of a kitchen infestation.
  • Insect growth regulator translocates deep into wall cavities, cracks and crevices.