Learn More
MSC Zoëcon Insecticide, 9% (S)-Hydroprene, 120 Day Control, Concentrate
Catalog No. 502836047
Encompass
This insect growth regulator features 9% (S)-Hydroprene, designed to break insect life cycles and provide 120 days of control. It utilizes a translocating active ingredient to reach insects in hard-to-treat areas like wall cavities, cracks, and crevices, and is approved for use in restaurant and other food handling establishments.
  • Features a translocating active ingredient to reach hard-to-treat areas.
  • Effectively breaks insects' life cycles.
  • Offers 110 days of control.
  • Juvenile hormone mimic prevents normal development.
  • Translocates deep into wall cavities, cracks, and crevices.
  • Approved for use in restaurant and other food handling establishments.
  • Can be used as a general surface spray, fogging, spot, or crack and crevice application.
Catalog No. 50-283-6047 Supplier MSC Supplier No. 74333394
This insect growth regulator features 9% (S)-Hydroprene, designed to break insect life cycles and provide 120 days of control. It utilizes a translocating active ingredient to reach insects in hard-to-treat areas like wall cavities, cracks, and crevices, and is approved for use in restaurant and other food handling establishments.
  • Features a translocating active ingredient to reach hard-to-treat areas.
  • Effectively breaks insects' life cycles.
  • Offers 110 days of control.
  • Juvenile hormone mimic prevents normal development.
  • Translocates deep into wall cavities, cracks, and crevices.
  • Approved for use in restaurant and other food handling establishments.
  • Can be used as a general surface spray, fogging, spot, or crack and crevice application.