Learn More
MSC Zoecon Aerosol, Dual-Action, Low Odor, 18 oz
Catalog No. 502839282
Encompass
Gentrol Complete Aerosol combines an insecticide and an insect growth regulator (IGR) to break the life cycles of cockroaches, fruit flies, drain flies, and more. It provides knockdown and kill with residual control to prevent future insect infestations, helping to save on labor costs.
  • Combines an insecticide and an insect growth regulator for comprehensive pest management.
  • Active ingredient disrupts insect life cycle, preventing rebound infestations.
  • Provides quick knockdown and lasting residual control.
  • Broad-spectrum product effective against many pests including cockroaches, fruit flies, and drain flies.
  • Approved for use in sensitive areas like hospitals and food establishments.
  • Low odor, ideal for indoor applications.
  • Convenient one-step application as a crack and crevice or spot treatment.
Catalog No. 50-283-9282 Supplier MSC Supplier No. 74323148
Gentrol Complete Aerosol combines an insecticide and an insect growth regulator (IGR) to break the life cycles of cockroaches, fruit flies, drain flies, and more. It provides knockdown and kill with residual control to prevent future insect infestations, helping to save on labor costs.
  • Combines an insecticide and an insect growth regulator for comprehensive pest management.
  • Active ingredient disrupts insect life cycle, preventing rebound infestations.
  • Provides quick knockdown and lasting residual control.
  • Broad-spectrum product effective against many pests including cockroaches, fruit flies, and drain flies.
  • Approved for use in sensitive areas like hospitals and food establishments.
  • Low odor, ideal for indoor applications.
  • Convenient one-step application as a crack and crevice or spot treatment.