Learn More
Enterprise Technology Solutions Genuine Joe Bottles, Plastic, 32oz, 12/CT, Translucent
SDP
Catalog No. 5000831123
Encompass_Preferred
Lightweight plastic bottle features convenient graduations for fast, accurate measurements. Translucent design lets you quickly see the amount of liquid that's remaining. Durable material is perfect for dispensing cleaners. 28mm/400 neck finish fits a variety of popular trigger sprayers (sold separately).
Catalog No. 50-008-31123 Supplier Enterprise Technology Solutions Supplier No. GJO85126
Lightweight plastic bottle features convenient graduations for fast, accurate measurements. Translucent design lets you quickly see the amount of liquid that's remaining. Durable material is perfect for dispensing cleaners. 28mm/400 neck finish fits a variety of popular trigger sprayers (sold separately).