Learn More
Enterprise Technology Solutions Genuine Joe Dish Detergent, Concentrated, Squeeze Bottle, 38 oz, Blue
SDP
Catalog No. 5000811028
Encompass_Preferred
Dishwashing detergent leaves dishes sparkling clean and film-free. It is ideal for cleaning heavily encrusted pots and pans and powers through the toughest grease. Premium Dish Detergent is specifically designed for use where fast grease emulsification and lasting suds are a must. It is recommended for supermarkets, institutional facilities and restaurants. Mountains of hardworking suds penetrate grease and food deposits leaving pots, pans, glassware and china spot-free. Concentrated, liquid formula offers a dilution ratio of 1:512 (1/4 oz. per gallon). Detergent comes in a squeeze bottle with a flip top for easy dispensing.
Catalog No. 50-008-11028 Supplier Enterprise Technology Solutions Supplier No. GJO99679
Dishwashing detergent leaves dishes sparkling clean and film-free. It is ideal for cleaning heavily encrusted pots and pans and powers through the toughest grease. Premium Dish Detergent is specifically designed for use where fast grease emulsification and lasting suds are a must. It is recommended for supermarkets, institutional facilities and restaurants. Mountains of hardworking suds penetrate grease and food deposits leaving pots, pans, glassware and china spot-free. Concentrated, liquid formula offers a dilution ratio of 1:512 (1/4 oz. per gallon). Detergent comes in a squeeze bottle with a flip top for easy dispensing.