Learn More
Enterprise Technology Solutions Genuine Joe Gloves,Disposable,PowderFree,Interior PU Coating,Vinyl,Small,3.5mil,100/BX,Clear
SDP
Catalog No. 5000827892
Encompass_Preferred
These Vinyl Disposable Gloves deliver the perfect balance of value, comfort and protection for everyday use. These cost-effective gloves are perfect for businesses and individuals requiring high-volume hand protection. The interior PU coating ensures they slide on and off effortlessly, keeping your hands comfortable during frequent changes. Powder-free material measures 3mil thick for added strength and durability.
Catalog No. 50-008-27892 Supplier Enterprise Technology Solutions Supplier No. GJO15432
These Vinyl Disposable Gloves deliver the perfect balance of value, comfort and protection for everyday use. These cost-effective gloves are perfect for businesses and individuals requiring high-volume hand protection. The interior PU coating ensures they slide on and off effortlessly, keeping your hands comfortable during frequent changes. Powder-free material measures 3mil thick for added strength and durability.