Learn More
Enterprise Technology Solutions Genuine Joe Gloves,Disposable,PowderFree,Tactile,PrecisionFit,Latex,Large,4.0mil,100/BX,White
SDP
Catalog No. 5000828338
Encompass_Preferred
These Professional-Grade Latex Gloves provide hand protection and safety without sacrificing comfort. Allergy-friendly gloves deliver the superior tactile sensitivity of natural rubber with the clean, non-irritating wear of a powder-free finish. Precision-fit gloves measure 4.0mil thick and offer a comfortable, secure fit with reliable grip and dexterity.
Catalog No. 50-008-28338 Supplier Enterprise Technology Solutions Supplier No. GJO15438
These Professional-Grade Latex Gloves provide hand protection and safety without sacrificing comfort. Allergy-friendly gloves deliver the superior tactile sensitivity of natural rubber with the clean, non-irritating wear of a powder-free finish. Precision-fit gloves measure 4.0mil thick and offer a comfortable, secure fit with reliable grip and dexterity.