Learn More
Enterprise Technology Solutions Genuine Joe Indoor Mat, Moisture Absorbent, Vinyl Back, 3'x5', Charcoal
SDP
Catalog No. 5000831122
Encompass_Preferred
Indoor mat features a plush surface that effectively absorbs moisture to help keep the surrounding areas clean and dry. Quick-drying material maintains its efficient performance for effective use in high-traffic locations. Durable vinyl backing resists sliding on smooth surfaces to prevent frequent adjustments. Floor mat is designed for indoor use.
Catalog No. 50-008-31122 Supplier Enterprise Technology Solutions Supplier No. GJO56352
Indoor mat features a plush surface that effectively absorbs moisture to help keep the surrounding areas clean and dry. Quick-drying material maintains its efficient performance for effective use in high-traffic locations. Durable vinyl backing resists sliding on smooth surfaces to prevent frequent adjustments. Floor mat is designed for indoor use.