Learn More
Enterprise Technology Solutions Genuine Joe Nitrile Gloves,Powder-Free,Latex-Free,Ambidextrous,BeadedCuff,XLarge,4.0mil,100/BX,Blue
SDP
Catalog No. 5000828134
Encompass_Preferred
Premium Nitrile Disposable Gloves deliver a soft, pliable fit with excellent tactile sensitivity. They are ideal for homecare, food-handling, housing and beauty industries. Textured fingers enhance your grip. Beaded cuff on these extra-large gloves allows a secure fit. Powder-free, latex-free nitrile is 4 mil thick. For easier access, professional-grade, nonsterile gloves are ambidextrous so you can use on either hand. They offer a shelf life of 5 years under dry storage conditions.
Catalog No. 50-008-28134 Supplier Enterprise Technology Solutions Supplier No. GJO15411
Premium Nitrile Disposable Gloves deliver a soft, pliable fit with excellent tactile sensitivity. They are ideal for homecare, food-handling, housing and beauty industries. Textured fingers enhance your grip. Beaded cuff on these extra-large gloves allows a secure fit. Powder-free, latex-free nitrile is 4 mil thick. For easier access, professional-grade, nonsterile gloves are ambidextrous so you can use on either hand. They offer a shelf life of 5 years under dry storage conditions.