Learn More
Enterprise Technology Solutions Genuine Joe Nitrile Gloves,Powder-Free,Latex-Free,NonSterile,Ambidextrous,BeadedCuff,Small,3.0mil,100/BX,Black
SDP
Catalog No. 5000827973
Encompass_Preferred
Premium Nitrile Disposable Gloves deliver a soft, pliable fit with excellent tactile sensitivity. Ambidextrous gloves have textured fingers for better grip and a beaded cuff for added durability. Powder-free, latex-free material measures 3.0mil thick and is suitable for homecare, food handling, IT, housing and beauty industries. Gloves have a shelf life of five years under dry storage conditions.
Catalog No. 50-008-27973 Supplier Enterprise Technology Solutions Supplier No. GJO15404
Premium Nitrile Disposable Gloves deliver a soft, pliable fit with excellent tactile sensitivity. Ambidextrous gloves have textured fingers for better grip and a beaded cuff for added durability. Powder-free, latex-free material measures 3.0mil thick and is suitable for homecare, food handling, IT, housing and beauty industries. Gloves have a shelf life of five years under dry storage conditions.