Learn More
Enterprise Technology Solutions Genuine Joe Nitrile Gloves,PowderFree,NonSterile,Ambidextrous,Textured.BeadedCuff,Medium,5.0mil,100/BX,Black
SDP
Catalog No. 5000827921
Encompass_Preferred
These Heavy-Duty Nitrile Gloves are engineered for durability and comfort. They offer exceptional protection without compromising flexibility. Powder-free, non-sterile gloves are designed with an ambidextrous fit for convenient versatile use. Finger-textured surface helps with grip, and beaded cuffs provide a more secure fit. Durable material measures 5.0mil thick for added strength. Gloves are ideal for high-performance tasks across a variety of industries.
Catalog No. 50-008-27921 Supplier Enterprise Technology Solutions Supplier No. GJO15421
These Heavy-Duty Nitrile Gloves are engineered for durability and comfort. They offer exceptional protection without compromising flexibility. Powder-free, non-sterile gloves are designed with an ambidextrous fit for convenient versatile use. Finger-textured surface helps with grip, and beaded cuffs provide a more secure fit. Durable material measures 5.0mil thick for added strength. Gloves are ideal for high-performance tasks across a variety of industries.