Learn More
Enterprise Technology Solutions Genuine Joe Nitrile Gloves,PowderFree,NonSterile,Ambidextrous,TexturedFinger,BeadedCuff,Large,5.0mil,100/BX,BE
SDP
Catalog No. 5000828135
Encompass_Preferred
High-quality Powder-Free Nitrile Gloves are designed for versatility with an ambidextrous fit. Textured finger surface provides better grip, while the beaded cuff ensures a secure fit. These soft-yet-durable gloves are ideal for a wide range of industries, including healthcare, lab work, foodservice, beauty and IT handling. Strong material measures 5.0mil thick and is powder-free. Non-sterile loves have a shelf life of five years under dry, dark storage
Catalog No. 50-008-28135 Supplier Enterprise Technology Solutions Supplier No. GJO15418
High-quality Powder-Free Nitrile Gloves are designed for versatility with an ambidextrous fit. Textured finger surface provides better grip, while the beaded cuff ensures a secure fit. These soft-yet-durable gloves are ideal for a wide range of industries, including healthcare, lab work, foodservice, beauty and IT handling. Strong material measures 5.0mil thick and is powder-free. Non-sterile loves have a shelf life of five years under dry, dark storage