Learn More
Enterprise Technology Solutions Genuine Joe Nylon Hair Net, Nonwoven, Large, 21", 100/PK,BK
SDP
Catalog No. 5000828313
Encompass_Preferred
Hair net keeps employees' hair in place to prevent accidental contamination. Honeycomb design lets air circulate for comfortable use during extended work periods. Lightweight nylon material features a durable construction to resist tearing. FDA-compliant hairnet is excellent for use in food processing, food handling and manufacturing facilities.
Catalog No. 50-008-28313 Supplier Enterprise Technology Solutions Supplier No. GJO85135
Hair net keeps employees' hair in place to prevent accidental contamination. Honeycomb design lets air circulate for comfortable use during extended work periods. Lightweight nylon material features a durable construction to resist tearing. FDA-compliant hairnet is excellent for use in food processing, food handling and manufacturing facilities.