Learn More
Enterprise Technology Solutions Genuine Joe Plastic Bottle, w/Graduation Marks, 24oz, 24/CT, Translucent
SDP
Supplier:  Enterprise Technology Solutions GJO85139
Encompass_Preferred
Plastic bottle with a 28mm/400 neck finish is made of polyethylene to safely contain a wide variety of liquids. Convenient graduation marks allow you to make quick measurements inside the bottle. Textured upper surface enhances your grip to provide a firm grip at all times. Clear, translucent bottle lets you easily monitor the current level of liquid contained.
Catalog No. 50-008-30716