Learn More
Enterprise Technology Solutions Genuine Joe Toilet Bowl Brush, Deluxe, PET Bristles, Blue/Gray
SDP
Catalog No. 5000830873
Encompass_Preferred
Toilet bowl brush with ergonomic comfort grip features an all-plastic construction to clean every area of your bowl without scratching like traditional wire brushes. Scratchless, chemical-resistant design works well with all types of cleaners to help you quickly power away stains. Durable bristles are made of PET. Handle is made of polypropylene and thermoplastic rubber.
Catalog No. 50-008-30873 Supplier Enterprise Technology Solutions Supplier No. GJO99687
Toilet bowl brush with ergonomic comfort grip features an all-plastic construction to clean every area of your bowl without scratching like traditional wire brushes. Scratchless, chemical-resistant design works well with all types of cleaners to help you quickly power away stains. Durable bristles are made of PET. Handle is made of polypropylene and thermoplastic rubber.