Learn More
Enterprise Technology Solutions Genuine Joe Trigger Sprayer, Chemical Resistant, 28mm, 24/CT, Yellow
SDP
Supplier:  Enterprise Technology Solutions GJO85145
Encompass_Preferred
Chemical-resistant trigger sprayer accurately dispenses liquids directly to surfaces. Adjustable nozzle allows you to quickly control the amount of liquid to be dispensed for economical use. Easy-to-use trigger allows fast, repeatable use for efficient performance. Sprayer is designed for bottles with a 28mm/400 neck finish (sold separately).
Catalog No. 50-008-30715