Learn More
Enterprise Technology Solutions Genuine Joe Trigger Sprayer, For Liquid Cleaners, Plastic, Gray
SDP
Catalog No. 5000831626
Encompass_Preferred
Trigger sprayer accurately dispenses liquids directly to surfaces. Adjustable nozzle allows you to quickly control the amount of liquid to be dispensed for economical use. Easy-to-use trigger allows fast, repeatable use for efficient performance. Sprayer with 10" dip tube is designed for bottles with a 28mm/400 neck finish (sold separately). It fits well in 32 oz. bottles.
Catalog No. 50-008-31626 Supplier Enterprise Technology Solutions Supplier No. GJO85119
Trigger sprayer accurately dispenses liquids directly to surfaces. Adjustable nozzle allows you to quickly control the amount of liquid to be dispensed for economical use. Easy-to-use trigger allows fast, repeatable use for efficient performance. Sprayer with 10" dip tube is designed for bottles with a 28mm/400 neck finish (sold separately). It fits well in 32 oz. bottles.