Learn More
Enterprise Technology Solutions Genuine Joe Trigger Sprayer, Standard, 14.25"x10.75"x4.13", 24/CT, RD/WE
SDP
Catalog No. 5000831217
Encompass_Preferred
General-purpose trigger sprayer helps you accurately dispense liquids directly to any area. Adjustable nozzle lets you easily control how much liquid you want to spray to prevent wasteful use. Easy-to-use trigger allows fast, repeatable use to maximize your efficiency. Sprayer attaches to quart-sized bottles with a 28mm/400 neck finish.
Catalog No. 50-008-31217 Supplier Enterprise Technology Solutions Supplier No. GJO85131
General-purpose trigger sprayer helps you accurately dispense liquids directly to any area. Adjustable nozzle lets you easily control how much liquid you want to spray to prevent wasteful use. Easy-to-use trigger allows fast, repeatable use to maximize your efficiency. Sprayer attaches to quart-sized bottles with a 28mm/400 neck finish.