Learn More
Abcam Genz-123346, 10MG
Supplier: Abcam AB28698610MG
MW 418.6 Da, Purity >=98%. Potent and selective glucosylceramide synthase inhibitor with potential anticancer activity. Does not affect the activities of other related glycosidase, such as 1-Oacylceramide synthase, a-glucosidase, and glucocerebrosidase in any significant manner. Shown to improve glucose tolerance, improve insulin sensitivity, and normalize hemoglobin A1C levels in Zucker diabetic fatty rats.
The product is subject to the following: Abcam Restricted Use Statement