Learn More
Cayman Chemical Genz-123346, Purity: ≥98%, CAS: 491833-30-8, 50 mg
SDP
Catalog No. 502940385
Encompass_Preferred
A glucosylceramide synthase inhibitor (IC50 = 14 nM); reduces hepatic levels of glucosylceramide and triglycerides as well as blood levels of ALT, hemoglobin A1c, and non-fasting glucose in ob/ob mice at 120 mg/kg per day; decreases hepatic fat mass and reduces the size and number of hepatic lipid droplets in diet-induced obese mice at 125 mg/kg per day
Catalog No. 50-294-0385 Supplier Cayman Chemical Supplier No. 2850050 MG
A glucosylceramide synthase inhibitor (IC50 = 14 nM); reduces hepatic levels of glucosylceramide and triglycerides as well as blood levels of ALT, hemoglobin A1c, and non-fasting glucose in ob/ob mice at 120 mg/kg per day; decreases hepatic fat mass and reduces the size and number of hepatic lipid droplets in diet-induced obese mice at 125 mg/kg per day