Learn More
Selleck Chemical LLC Genz-123346 free base
SDP
Supplier:  Selleck Chemical LLC S64965MG
Encompass_Preferred
Genz-123346 is an inhibitor of GL1 synthase that blocks the conversion of ceramide to GL1 the first step in the biosynthesis of gangliosides and other glycosphingolipids
Catalog No. 50-000-02984