Learn More
Medchemexpress LLC HY-16228 5mg Medchemexpress, Genz-644282 CAS:529488-28-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY162285MG
Encompass_Preferred
Medchemexpress, HY-16228 5mg Genz-644282 CAS:529488-28-6 Genz-644282 is a non-camptothecin topoisomerase I inhibitor, used for cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0709