Learn More
Mesa Laboratories GEOBACILLUS STEAROTHERMOPHILUS
SDP
Catalog No. NC1523615
Encompass
Geobacillus stearothermophilus spore suspension in aqueous solution 10e7/0.1mL - SSA/7
This item is not returnable. View return policy
Catalog No. NC1523615 Supplier Mesa Laboratories Supplier No. SSSA/7
 Restricted Item
Add to Cart
Add to Cart
Geobacillus stearothermophilus spore suspension in aqueous solution 10e7/0.1mL - SSA/7