Learn More
ATCC Geobacillus stearothermophilus N
Catalog No. 502382139
Encompass
This strain is recommended by ATCC for use in the tests described in AOAC Official Methods 982.15, 982.18, and 988.08 where only the taxon is specified. Used in the assay of beta-lactam antibiotics.
Catalog No. 50-238-2139 Supplier ATCC Supplier No. 10149
This strain is recommended by ATCC for use in the tests described in AOAC Official Methods 982.15, 982.18, and 988.08 where only the taxon is specified. Used in the assay of beta-lactam antibiotics.