Learn More
ATCC Desulfovibrio vulgaris
Catalog No. 502381339
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Desulfovibrio vulgaris strain C-6 [CT1, IFO 13699, NCIB 8372] is a whole-genome sequenced strain. This anaerobic bacterium is cited to produce D(-)-lactate dehydrogenase and it has applications in bioresistance testing.
Catalog No. 50-238-1339 Supplier ATCC Supplier No. 7757
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Desulfovibrio vulgaris strain C-6 [CT1, IFO 13699, NCIB 8372] is a whole-genome sequenced strain. This anaerobic bacterium is cited to produce D(-)-lactate dehydrogenase and it has applications in bioresistance testing.