Learn More
ATCC Geobacillus stearothermophilus
Catalog No. 502384256
Encompass
This strain is recommended by ATCC for use in the tests described in military specification MIL S-36586A and U.S. Pharmacopeia, 23rd rev., <1211>, pp. 1976-1981, 1995 where only the taxon is specified. Steam and gas sterilization control.
Catalog No. 50-238-4256 Supplier ATCC Supplier No. 7953
This strain is recommended by ATCC for use in the tests described in military specification MIL S-36586A and U.S. Pharmacopeia, 23rd rev., <1211>, pp. 1976-1981, 1995 where only the taxon is specified. Steam and gas sterilization control.