Learn More
ATCC Geobacillus stearothermophilus
Supplier:  ATCC 12980
Encompass
Geobacillus stearothermophilus strain NCA 26 is a whole-genome sequenced bacterial type strain. The culture produces the restriction endonuclease BstPI and is used as a steam sterilization control and sterility assurance organism.
Catalog No. 50-238-2995