Learn More
ATCC Burkholderia humptydooensis; Strain: MSMB43
Catalog No. 502380038
Encompass
Type strain. Depositor claims resistant to amoxicillin / clavulanic acid, gentamicin, and streptomycin.
Catalog No. 50-238-0038 Supplier ATCC Supplier No. BAA2767
Type strain. Depositor claims resistant to amoxicillin / clavulanic acid, gentamicin, and streptomycin.