Learn More
ATCC Geobacter sulfurreducens; Strain PCA
Supplier:  ATCC 51573
Encompass
Geobacter sulfurreducens strain PCA was isolated from surface sediments in Norman, Oklahoma. This whole-genome sequenced bacterial type strain can be used in biotechnology.
Catalog No. 50-238-2157