Learn More
AdipoGen Geodin (5 mg)
SDP
Catalog No. 502053516
Encompass_Preferred
Chemical. CAS: 427-63-4. Formula: C17H12Cl2O7. MW: 399.2. Isolated from Penicillium sp. FKI-5002. Plasminogen activator inhibitor (PAI-1). Glucose uptake stimulator.
Catalog No. 50-205-3516 Supplier AdipoGen Supplier No. AGCN20139M005
Chemical. CAS: 427-63-4. Formula: C17H12Cl2O7. MW: 399.2. Isolated from Penicillium sp. FKI-5002. Plasminogen activator inhibitor (PAI-1). Glucose uptake stimulator.