Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Phoenix™ Geologic Journeys Deluxe Kit Components
SCI_ED

Catalog No. S395554GJ Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Plastic Container
Plastic bag
Plastic cup
Spray bottle
White flour
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S395554GJ White flour
S395584GJ Plastic cup
S395594GJ Spray bottle
S395604GJ Plastic bag
S395634GJ Plastic Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S395554GJ
Add to Cart
Add to Cart

Replacement parts for Educational Kit

Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.

Specifications

Grade K to 5
For Use With (Application) Education Kit
Quantity 5 lb. Bag
Product Type White flour
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.