Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Geology Time Scale Poster
GREENER_CHOICESCI_ED

Catalog No. s42871 Shop All Aldon Products
Change view
Click to view available options
Dimensions (L x W):
11 x 17 in.
21 x 34 in.
Product Type:
100# Paper Poster
Laminated Poster
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Product Type
S42871 21 x 34 in. 100# Paper Poster
S42870 11 x 17 in. Laminated Poster
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S42871 Supplier Innovating Science™ Supplier No. IS8751
Only null left
Add to Cart
Add to Cart

Ideal visual aid for teaching geology events. Innovating Science™ Geology Time Scale Poster shows major evolutionary events from 4.5 Billion Years Ago to present.

  • Students can visualize the milestones of geologic history
  • The events are color coded to the Period or Epoch in which they occurred

Specifications

Color White
Grade 8 to 9
Age 13 to 20
Dimensions (L x W) 21 x 34 in.
For Use With (Application) Geological time scale
Includes Geology poster
Quantity 1
Weight (English) 0.7 lb.
Class Size 1 Class
Product Type 100# Paper Poster
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.