Learn More
Geotech Environmental Equiptment Inc Geopump II Peristaltic Sampling Pump w/ Pump Head‚ AC/DC Power Accessories & Case
SDP
Catalog No. NC1817407
Encompass_Preferred
Geopump Series II portable sampling pumps offer reliable pressure or vacuum pumping of liquids. Peristaltic operation ensures the sample only comes in contact with the tubing‚ allowing for consistent sample integrity and easy cleaning. Use for field sampling from shallow wells and all surface water sources‚ as well as for laboratory applications.
Catalog No. NC1817407 Supplier Geotech Environmental Equiptment Inc Supplier No. 91352123
Geopump Series II portable sampling pumps offer reliable pressure or vacuum pumping of liquids. Peristaltic operation ensures the sample only comes in contact with the tubing‚ allowing for consistent sample integrity and easy cleaning. Use for field sampling from shallow wells and all surface water sources‚ as well as for laboratory applications.