Learn More
Enterprise Technology Solutions Georgia-Pacific HandSoap,Refill,Foam,Aloe,f/CounterMountDisp,1800mL,2/CT,GN
SDP
Catalog No. 5000831204
Encompass_Preferred
enMotion Gentle Foam Soap with Moisturizers features a rich foaming lather and mild conditioning formula that leaves skin feeling fresh and is gentle enough for daily use. Closed hygienic refill bottles install with one hand and dispense at least 99-percent of foam product before replacement is needed. Use with compatible enMotion Automated Touchless Counter Mount Soap Dispenser (52065) (not included).
Catalog No. 50-008-31204 Supplier Enterprise Technology Solutions Supplier No. GPC42718
enMotion Gentle Foam Soap with Moisturizers features a rich foaming lather and mild conditioning formula that leaves skin feeling fresh and is gentle enough for daily use. Closed hygienic refill bottles install with one hand and dispense at least 99-percent of foam product before replacement is needed. Use with compatible enMotion Automated Touchless Counter Mount Soap Dispenser (52065) (not included).