Learn More
Enterprise Technology Solutions Georgia-Pacific Shop Towels,Heavy-duty,9-1/10"x16-1/2",100/BX,White
SDP
Catalog No. 5000811186
Encompass_Preferred
Premium, folded wipers come in convenient dispenser boxes. Multiuse wiper (ultimate wiper to replace rags) is low lint, chemical-resistant and abrasion-resistant while offering a good absorption rate. Wipers are good with oils and water-based solvents since there are no binders used.
Catalog No. 50-008-11186 Supplier Enterprise Technology Solutions Supplier No. GPC25070
Premium, folded wipers come in convenient dispenser boxes. Multiuse wiper (ultimate wiper to replace rags) is low lint, chemical-resistant and abrasion-resistant while offering a good absorption rate. Wipers are good with oils and water-based solvents since there are no binders used.