Learn More
Description
- Explore the amazing world of ants up close with the GeoSafari Ant Factory, with sand
- Set includes everything kids need to raise and observe an ant colony, including an escape-proof ant habitat with large viewing area, magnification spots, and stand; special ant sand; feeding tool, and certificate to receive live ants.
- Settle your ants and watch as they work together to tunnel, eat, sleep, and more. Perfect for young entomologists; provides a 360° view of one of the world's most industrious insects
- Includes activity guide and mail-in certificate for Harvester Ants
- $4.00 shipping and handling not included
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 1 Class |
| Product Type | GeoSafari Ant Factories |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
